2-Chloro-4-fluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22438G100 AS22438G25
ACTIAS22438G100EA 0 GBP
ACTIAS22438G100 ACTIAS22438G25
2-Chloro-4-fluorobenzonitrile
2-Chloro-4-fluorobenzonitrile
Formula: C₇H₃ClFN
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 63…65 °C
MDL Number: MFCD00042523
CAS Number: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR