3-Chloro-2-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21651G100 AS21651G25
ACTIAS21651G100EA 0 GBP
ACTIAS21651G100 ACTIAS21651G25
3-Chloro-2-nitrobenzoic acid
3-Chloro-2-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Melting Pt: 236…240 °C
Density: 1,566 g/cm³ (18 °C)
MDL Number: MFCD00007087
CAS Number: 4771-47-5
EINECS: 225-313-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR