2-Chloro-6-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20460G100 AS20460G25 AS20460G5
ACTIAS20460G100EA 78 GBP
ACTIAS20460G100 ACTIAS20460G25 ACTIAS20460G5
2-Chloro-6-nitrobenzoic acid
2-Chloro-6-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD00100425
CAS Number: 5344-49-0
EINECS: 226-287-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR