2-Chloro-3-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20583G25 AS20583G5
ACTIAS20583G25EA 0 GBP
ACTIAS20583G25 ACTIAS20583G5
2-Chloro-3-nitrobenzoic acid
2-Chloro-3-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS Number: 3970-35-2
EINECS: 223-590-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR