4-Amino-2-chlorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20071G100 AS20071G25
ACTIAS20071G100EA 0 GBP
ACTIAS20071G100 ACTIAS20071G25
4-Amino-2-chlorobenzoic acid
4-Amino-2-chlorobenzoic acid
Formula: C₇H₆ClNO₂
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007772
CAS Number: 2457-76-3
EINECS: 219-540-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR