3,5-Dichlorobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20245G100
AS20245G25
ACTIAS20245G100EA
0
GBP
ACTIAS20245G100
ACTIAS20245G25
3,5-Dichlorobenzonitrile
3,5-Dichlorobenzonitrile
Formula:
C₇H₃Cl₂N Boiling Pt: 269 °C (1013 hPa) Melting Pt: 66 °C |
MDL Number:
MFCD00001800 CAS Number: 6575-00-4 EINECS: 229-495-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...