5-Chloro-2-nitrobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21855G100 AS21855G25 AS21855G5
ACTIAS21855G100EA 0 GBP
ACTIAS21855G100 ACTIAS21855G25 ACTIAS21855G5
5-Chloro-2-nitrobenzaldehyde
5-Chloro-2-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 77.5
Storage Temperature: Ambient
MDL Number: MFCD00007289
CAS Number: 6628-86-0
EINECS: 229-614-9
UN: 3077
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR