2-Chloro-5-nitrobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20316G100 AS20316G25
ACTIAS20316G100EA 0 GBP
ACTIAS20316G100 ACTIAS20316G25
2-Chloro-5-nitrobenzaldehyde
2-Chloro-5-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS Number: 6361-21-3
EINECS: 228-830-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR