5-Chlorosalicylaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20797G100
AS20797G25
ACTIAS20797G100EA
0
GBP
ACTIAS20797G100
ACTIAS20797G25
5-Chlorosalicylaldehyde
5-Chlorosalicylaldehyde
Formula:
C₇H₅ClO₂ Boiling Pt: 105 °C (12 mmHg) Melting Pt: 100.3 °C |
MDL Number:
MFCD00003331 CAS Number: 635-93-8 EINECS: 211-244-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...