2,5-Dichlorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS21515G100 AS21515G25
ACTIAS21515G100EA 113 GBP
ACTIAS21515G100 ACTIAS21515G25
2,5-Dichlorobenzaldehyde
2,5-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
MDL Number: MFCD00156140
CAS Number: 6361-23-5
EINECS: 228-832-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR