5-Methoxysalicylic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Hydroxy-5-methoxybenzoic acid
AS20849G100
AS20849G25
ACTIAS20849G100EA
0
GBP
ACTIAS20849G100
ACTIAS20849G25
5-Methoxysalicylic acid
5-Methoxysalicylic acid
Formula:
C₈H₈O₄ MW: 168.15 g/mol Boiling Pt: 327 °C (1013 hPa) Melting Pt: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002459 CAS Number: 2612-02-4 EINECS: 220-037-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...