3,4-Dinitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20458G100 AS20458G25
ACTIAS20458G100EA 0 GBP
ACTIAS20458G100 ACTIAS20458G25
3,4-Dinitrobenzoic acid
3,4-Dinitrobenzoic acid
Formula: C₇H₄N₂O₆
Melting Pt: 165.5…166.5 °C
Density: 1.674 g/cm³ (20 °C)
MDL Number: MFCD00007255
CAS Number: 528-45-0
EINECS: 208-433-9
Merck Index: 13,03303

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR