3-Hydroxy-4-nitrobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22000G100 AS22000G25
ACTIAS22000G100EA 0 GBP
ACTIAS22000G100 ACTIAS22000G25
3-Hydroxy-4-nitrobenzoic acid
3-Hydroxy-4-nitrobenzoic acid
Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 234…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00007110
CAS Number: 619-14-7
EINECS: 210-580-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR