4,5-Dimethoxyanthranilic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Amino-4,5-dimethoxybenzoic acid, 6-Aminoveratric acid

AS20066G100 AS20066G25
ACTIAS20066G100EA 75 GBP
ACTIAS20066G100 ACTIAS20066G25
4,5-Dimethoxyanthranilic acid
4,5-Dimethoxyanthranilic acid
Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 155…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00011671
CAS Number: 5653-40-7
EINECS: 227-095-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR