4-Cyanobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20131G100 AS20131G25
ACTIAS20131G100EA 0 GBP
ACTIAS20131G100 ACTIAS20131G25
4-Cyanobenzoic acid
4-Cyanobenzoic acid
Formula: C₈H₅NO₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 220…222 °C
Density: 1,382 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002528
CAS Number: 619-65-8
EINECS: 210-606-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR