3-Cyanobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS23348G100
AS23348G25
ACTIAS23348G100EA
135
GBP
ACTIAS23348G100
ACTIAS23348G25
3-Cyanobenzoic acid
3-Cyanobenzoic acid
Formula:
C₈H₅NO₂ MW: 147.13 g/mol Melting Pt: 219 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002486 CAS Number: 1877-72-1 EINECS: 217-511-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...