3-Hydroxybenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

AS22969G100 AS22969G25
ACTIAS22969G100EA 202 GBP
ACTIAS22969G100 ACTIAS22969G25
3-Hydroxybenzonitrile
3-Hydroxybenzonitrile
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 84.5…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00002252
CAS Number: 873-62-1
EINECS: 212-845-4
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR