3-Aminobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Cyanoaniline, m-Aminobenzonitrile, m-Cyanoaniline

AS20068G100 AS20068G25
ACTIAS20068G100EA 108 GBP
ACTIAS20068G100 ACTIAS20068G25
3-Aminobenzonitrile
3-Aminobenzonitrile
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 288…290 °C (1013 hPa)
Melting Pt: 48…50 °C
Density: 1.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007756
CAS Number: 2237-30-1
EINECS: 218-800-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR