2-Phenoxybenzaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20691G25
AS20691G5
ACTIAS20691G25EA
0
GBP
ACTIAS20691G25
ACTIAS20691G5
2-Phenoxybenzaldehyde
2-Phenoxybenzaldehyde
Formula:
C₁₃H₁₀O₂ MW: 198.22 g/mol |
MDL Number:
MFCD00800666 CAS Number: 19434-34-5 EINECS: 243-061-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...