4-Methoxysalicylaldehyde
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Hydroxy-4-methoxybenzaldehyde
AS23889G100
AS23889G25
ACTIAS23889G100EA
0
GBP
ACTIAS23889G100
ACTIAS23889G25
4-Methoxysalicylaldehyde
4-Methoxysalicylaldehyde
Formula:
C₈H₈O₃ Boiling Pt: 124 °C (12 mmHg) Melting Pt: 42…43 °C Density: 1.2065 g/cm³ (51 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003327 CAS Number: 673-22-3 EINECS: 211-604-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...