4-Hydroxy-3-nitrobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20857G100 AS20857G25
ACTIAS20857G100EA 0 GBP
ACTIAS20857G100 ACTIAS20857G25
4-Hydroxy-3-nitrobenzaldehyde
4-Hydroxy-3-nitrobenzaldehyde
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00007117
CAS Number: 3011-34-5
EINECS: 221-141-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR