1-Bromo-3-iodobenzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3-Bromoiodobenzene

AS21709G100 AS21709G25
ACTIAS21709G100EA 89 GBP
ACTIAS21709G100 ACTIAS21709G25
1-Bromo-3-iodobenzene
1-Bromo-3-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: –9,3 °C
Density: 2,21 g/cm³ (20 °C)
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR