4-Bromo-2-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20029G100 AS20029G25
ACTIAS20029G100EA 0 GBP
ACTIAS20029G100 ACTIAS20029G25
4-Bromo-2-fluorobenzoic acid
4-Bromo-2-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00143263
CAS Number: 112704-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR