3-Bromo-4-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20539G100 AS20539G25
ACTIAS20539G100EA 0 GBP
ACTIAS20539G100 ACTIAS20539G25
3-Bromo-4-fluorobenzoic acid
3-Bromo-4-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00042463
CAS Number: 1007-16-5
EINECS: 213-751-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR