2-Bromo-4-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS23248G100 AS23248G25
ACTIAS23248G100EA 115 GBP
ACTIAS23248G100 ACTIAS23248G25
2-Bromo-4-fluorobenzoic acid
2-Bromo-4-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 173…175 °C
MDL Number: MFCD00055370
CAS Number: 1006-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR