2-Bromo-4-fluorobenzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22194G100 AS22194G25
ACTIAS22194G100EA 0 GBP
ACTIAS22194G100 ACTIAS22194G25
2-Bromo-4-fluorobenzonitrile
2-Bromo-4-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
MDL Number: MFCD00672924
CAS Number: 36282-26-5
EINECS: 252-949-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR