5-Bromo-2-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20644G100 AS20644G25
ACTIAS20644G100EA 0 GBP
ACTIAS20644G100 ACTIAS20644G25
5-Bromo-2-fluorobenzaldehyde
5-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
Boiling Pt: 229
Melting Pt: 49
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00070755
CAS Number: 93777-26-5
EINECS: 298-056-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR