2-Bromo-6-iodopyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS23918G1
AS23918G5
ACTIAS23918G1EA
0
GBP
ACTIAS23918G1
ACTIAS23918G5
2-Bromo-6-iodopyridine
2-Bromo-6-iodopyridine
Formula:
C₅H₃BrIN |
MDL Number:
MFCD08059557 CAS Number: 234111-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...