2-Amino-3-benzyloxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Benzyloxypyridin-2-amine, 3-Benzyloxy-2-pyridylamine

AS21692G100 AS21692G25
ACTIAS21692G100EA 114 GBP
ACTIAS21692G100 ACTIAS21692G25
2-Amino-3-benzyloxypyridine
2-Amino-3-benzyloxypyridine
Formula: C₁₂H₁₂N₂O
MW: 200.24 g/mol
MDL Number: MFCD00006316
CAS Number: 24016-03-3
EINECS: 245-983-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR