2,4-Difluorophenylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21502G100 AS21502G25
ACTIAS21502G100EA 0 GBP
ACTIAS21502G100 ACTIAS21502G25
2,4-Difluorophenylhydrazinium chloride
2,4-Difluorophenylhydrazinium chloride
Formula: C₆H₇ClF₂N₂
MDL Number: MFCD00013383
CAS Number: 51523-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR