4-Chlorophenylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21056G100 AS21056G25
ACTIAS21056G100EA 0 GBP
ACTIAS21056G100 ACTIAS21056G25
4-Chlorophenylhydrazinium chloride
4-Chlorophenylhydrazinium chloride
Formula: C₆H₈Cl₂N₂
MDL Number: MFCD00012943
CAS Number: 1073-70-7
EINECS: 214-030-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR