2,5-Dichlorophenylhydrazinium chloride

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21503G100 AS21503G25
ACTIAS21503G100EA 0 GBP
ACTIAS21503G100 ACTIAS21503G25
2,5-Dichlorophenylhydrazinium chloride
2,5-Dichlorophenylhydrazinium chloride
Formula: C₆H₇Cl₃N₂
MDL Number: MFCD00052266
CAS Number: 50709-35-8
EINECS: 256-729-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR