4-Fluorophenyl methyl sulphone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Fluoro-4-(methylsulphonyl)benzene, 1-Fluor-4-(methylsulphonyl)benzol

AS24103G100 AS24103G25
ACTIAS24103G100EA 0 GBP
ACTIAS24103G100 ACTIAS24103G25
4-Fluorophenyl methyl sulphone
4-Fluorophenyl methyl sulfone
Formula: C₇H₇FO₂S
MDL Number: MFCD00039753
CAS Number: 455-15-2
EINECS: 207-237-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR