4-Bromo-3-chloroaniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS33305G100 AS33305G25
ACTIAS33305G100EA 0 GBP
ACTIAS33305G100 ACTIAS33305G25
4-Bromo-3-chloroaniline
4-Bromo-3-chloroaniline
Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 67…68 °C
MDL Number: MFCD00041311
CAS Number: 21402-26-6
EINECS: 244-370-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR