5-Bromoanthranilic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS20090G100
AS20090G25
ACTIAS20090G100EA
0
GBP
ACTIAS20090G100
ACTIAS20090G25
5-Bromoanthranilic acid
5-Bromoanthranilic acid
Formula:
C₇H₆BrNO₂ Boiling Pt: 342 °C (1013 hPa) Melting Pt: 218…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007823 CAS Number: 5794-88-7 EINECS: 227-338-3 UN: 2811 ADR: 6.1,III Merck Index: 13,01389 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...