3,5-Dibromobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22318G100 AS22318G25
ACTIAS22318G100EA 0 GBP
ACTIAS22318G100 ACTIAS22318G25
3,5-Dibromobenzoic acid
3,5-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00051758
CAS Number: 618-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR