3-Bromobenzonitrile
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
m-Bromobenzonitrile
AS20103G100
AS20103G25
ACTIAS20103G100EA
0
GBP
ACTIAS20103G100
ACTIAS20103G25
3-Bromobenzonitrile
3-Bromobenzonitrile
Formula:
C₇H₄BrN Boiling Pt: 226 °C (1013 hPa) Melting Pt: 37…40 °C Density: 1.562 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001796 CAS Number: 6952-59-6 EINECS: 230-127-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...