3-Hydroxy-2-nitrobenzoic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21650G1
AS21650G5
ACTIAS21650G1EA
0
GBP
ACTIAS21650G1
ACTIAS21650G5
3-Hydroxy-2-nitrobenzoic acid
3-Hydroxy-2-nitrobenzoic acid
Formula:
C₇H₅NO₅ |
MDL Number:
MFCD00017003 CAS Number: 602-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...