(S)-(-)-3-Amino-1,2-propanediol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-3-Amino-1,2-propanediol, (S)-3-Aminopropane-1,2-diol
AS21336G25
AS21336G5
ACTIAS21336G25EA
0
GBP
ACTIAS21336G25
ACTIAS21336G5
(S)-(-)-3-Amino-1,2-propanediol
(S)-(-)-3-Amino-1,2-propanediol
Formula:
C₃H₉NO₂ MW: 91.11 g/mol |
MDL Number:
MFCD00798260 CAS Number: 61278-21-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...