(R)-(+)-3-Amino-1,2-propanediol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-3-Aminopropane-1,2-diol, (R)-3-Amino-1,2-propanediol

AS22830G100 AS22830G25 AS22830G5
ACTIAS22830G100EA 455 GBP
ACTIAS22830G100 ACTIAS22830G25 ACTIAS22830G5
(R)-(+)-3-Amino-1,2-propanediol
(R)-(+)-3-Amino-1,2-propanediol
Formula: C₃H₉NO₂
MW: 91.11 g/mol
MDL Number: MFCD00798261
CAS Number: 66211-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR