9,10-Diphenylanthracene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24436G100
AS24436G25
ACTIAS24436G100EA
0
GBP
ACTIAS24436G100
ACTIAS24436G25
9,10-Diphenylanthracene
9,10-Diphenylanthracene
Formula:
C₂₆H₁₈ Boiling Pt: 485 °C (1013 hPa) Melting Pt: 245 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001253 CAS Number: 1499-10-1 EINECS: 216-105-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...