6-Bromo-1-hexene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS21023G100 AS21023G25
ACTIAS21023G100EA 69 GBP
ACTIAS21023G100 ACTIAS21023G25
6-Bromo-1-hexene
6-Bromo-1-hexene
Formula: C₆H₁₁Br
MW: 163.06 g/mol
Boiling Pt: 34…36 °C (0.05 mmHg)
Density: 1.2169 g/cm³ (20 °C)
MDL Number: MFCD00000269
CAS Number: 2695-47-8
EINECS: 220-267-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR