ω-Bromo-4-hydroxyacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromo-1-(4-hydroxyphenyl)ethanone, 2-Bromo-4'-hydroxyacetophenone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, α-Bromo-4-hydroxyacetophenone, 4-Hydroxyphenacyl bromide
AS20735G100
AS20735G25
ACTIAS20735G100EA
0
GBP
ACTIAS20735G100
ACTIAS20735G25
ω-Bromo-4-hydroxyacetophenone
ω-Bromo-4-hydroxyacetophenone
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol |
MDL Number:
MFCD00072424 CAS Number: 2491-38-5 EINECS: 219-655-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...