Tributylallyltin

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51607G100 AS51607G25 AS51607G5
ACTIAS51607G100EA 0 GBP
ACTIAS51607G100 ACTIAS51607G25 ACTIAS51607G5
Tributylallyltin
Tributylallyltin
Formula: C₁₅H₃₂Sn
Boiling Pt: 88 °C (1013 hPa)
Density: 1.073 g/cm³ (25 °C)
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR