Triethyl orthobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: alpha,alpha,alpha-Triethoxytoluene, (Triethoxymethyl)benzene, Orthobenzoic acid triethyl ester

AS20921G100 AS20921G25
ACTIAS20921G100EA 0 GBP
ACTIAS20921G100 ACTIAS20921G25
Triethyl orthobenzoate
Triethyl orthobenzoate
Formula: C₁₃H₂₀O₃
MDL Number: MFCD00009222
CAS Number: 1663-61-2
EINECS: 216-771-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR