2-Hydroxy-5-nitro-4-picoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Hydroxy-4-methyl-5-nitropyridine, 4-Methyl-5-nitro-2-pyridone
AS21281G25
AS21281G5
ACTIAS21281G25EA
0
GBP
ACTIAS21281G25
ACTIAS21281G5
2-Hydroxy-5-nitro-4-picoline
2-Hydroxy-5-nitro-4-picoline
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol Melting Pt: 189…190 °C |
MDL Number:
MFCD00010690 CAS Number: 21901-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...