2-Hydroxy-5-nitro-3-picoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine
AS21310G100
AS21310G25
ACTIAS21310G100EA
191
GBP
ACTIAS21310G100
ACTIAS21310G25
2-Hydroxy-5-nitro-3-picoline
2-Hydroxy-5-nitro-3-picoline
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol |
MDL Number:
MFCD03095073 CAS Number: 21901-34-8 EINECS: 244-647-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...