2-Amino-5-nitro-4-picoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21280G100 AS21280G25 AS21280G5
ACTIAS21280G100EA 0 GBP
ACTIAS21280G100 ACTIAS21280G25 ACTIAS21280G5
2-Amino-5-nitro-4-picoline
2-Amino-5-nitro-4-picoline
Formula: C₆H₇N₃O₂
Melting Pt: 220 °C
MDL Number: MFCD00010692
CAS Number: 21901-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR