4-(Trifluoromethoxy)benzenesulphonamide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24811G100 AS24811G25
ACTIAS24811G100EA 0 GBP
ACTIAS24811G100 ACTIAS24811G25
4-(Trifluoromethoxy)benzenesulphonamide
4-(Trifluoromethoxy)benzenesulphonamide
Formula: C₇H₆F₃NO₃S
MDL Number: MFCD00798381
CAS Number: 1513-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR