2-(Trifluoromethoxy)benzenesulphonamide
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS25372G100
AS25372G25
AS25372G5
ACTIAS25372G100EA
0
GBP
ACTIAS25372G100
ACTIAS25372G25
ACTIAS25372G5
2-(Trifluoromethoxy)benzenesulphonamide
2-(Trifluoromethoxy)benzenesulphonamide
Formula:
C₇H₆F₃NO₃S MW: 241.19 g/mol |
MDL Number:
MFCD01320751 CAS Number: 37526-59-3 EINECS: 448-450-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...